AMBH324A5446 Ambeed, Inc.

(1S,4R)-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid

  • MDL number MFCD00211287
  • Cas Number : 151907-79-8
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/151907-79-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/151907-79-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart