AMBH324A5447 Ambeed, Inc.

(R)-2-(4-Hydroxy-6-methylnicotinamido)-2-(4-hydroxyphenyl)acetic acid

  • MDL number MFCD09033158
  • Cas Number : 70785-61-4
  • Molecular Weight : 302.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/70785-61-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/70785-61-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart