AMBH324A5455 Ambeed, Inc.

1-(2,4-Dihydroxy-6-methoxyphenyl)-3-phenylprop-2-en-1-one

  • MDL number MFCD00238554
  • Cas Number : 18956-16-6
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/18956-16-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/18956-16-6.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart