AMBH324A545E Ambeed, Inc.

(R)-1-(7-Bromoquinolin-2-yl)ethanol

  • MDL number MFCD29920821
  • Cas Number : 1509899-61-9
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1509899-61-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1509899-61-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart