AMBH324A5462 Ambeed, Inc.

Fmoc-Asp(OMpe)-OH

  • MDL number MFCD10001333
  • Cas Number : 180675-08-5
  • Molecular Weight : 439.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/180675-08-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/180675-08-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart