AMBH324A5464 Ambeed, Inc.

(R)-2-((4-Nitrophenethyl)amino)-1-phenylethanol hydrochloride

  • MDL number MFCD26407510
  • Cas Number : 521284-21-9
  • Molecular Weight : 322.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/521284-21-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/521284-21-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart