AMBH324A546E Ambeed, Inc.

(S)-1-tert-Butyl 2-methyl 4-methylenepyrrolidine-1,2-dicarboxylate

  • MDL number MFCD15832853
  • Cas Number : 84348-39-0
  • Molecular Weight : 241.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/84348-39-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/84348-39-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart