AMBH324A5478 Ambeed, Inc.

Didanosine

  • MDL number MFCD00077728
  • Cas Number : 69655-05-6
  • Molecular Weight : 236.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/69655-05-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69655-05-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart