AMBH324A547B Ambeed, Inc.

Acetyl tetrapeptide-9(Dermican LS 9837)

  • MDL number N/A
  • Cas Number : 928006-50-2
  • Molecular Weight : 539.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/928006-50-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/928006-50-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart