AMBH324A547C Ambeed, Inc.

2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol

  • MDL number MFCD00002244
  • Cas Number : 94-93-9
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/94-93-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/94-93-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart