AMBH324A5483 Ambeed, Inc.

(S)-2-Amino-3-(3-aminophenyl)propanoic acid dihydrochloride

  • MDL number MFCD29919675
  • Cas Number : 908571-75-5
  • Molecular Weight : 253.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/908571-75-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/908571-75-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart