AMBH324A5492 Ambeed, Inc.

(S)-1-(2-Methoxypyridin-4-yl)ethanamine hydrochloride

  • MDL number MFCD29044870
  • Cas Number : 1914157-93-9
  • Molecular Weight : 188.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1914157-93-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1914157-93-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart