AMBH324A549A Ambeed, Inc.

3,4,5-Trifluoro-4'-((trans,trans)-4'-propyl-[1,1'-bi(cyclohexan)]-4-yl)-1,1'-biphenyl

  • MDL number MFCD13188632
  • Cas Number : 137529-41-0
  • Molecular Weight : 414.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/137529-41-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/137529-41-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart