AMBH324A54A4 Ambeed, Inc.

1-((2S,3S)-2-(Benzyloxy)pentan-3-yl)-4-(4-(4-(4-hydroxyphenyl)piperazin-1-yl)phenyl)-1H-1,2,4-triazol-5(4H)-one

  • MDL number MFCD11977288
  • Cas Number : 184177-83-1
  • Molecular Weight : 513.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/184177-83-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/184177-83-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart