AMBH324A54A5 Ambeed, Inc.

(E)-1-Methyl-4-(2-nitrovinyl)benzene

  • MDL number MFCD00024828
  • Cas Number : 5153-68-4
  • Molecular Weight : 163.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5153-68-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5153-68-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart