AMBH324A54B2 Ambeed, Inc.

(Trans,trans)-4'-propyl[1,1'-Bicyclohexyl]-4-carboxyllic acid-4-(trans-4-propylcyclohexyl)phenyl ester

  • MDL number MFCD00673705
  • Cas Number : 88038-92-0
  • Molecular Weight : 452.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/88038-92-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/88038-92-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart