AMBH324A54B3 Ambeed, Inc.

H-D-Asn(Trt)-OH

  • MDL number MFCD00236802
  • Cas Number : 200192-49-0
  • Molecular Weight : 374.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/200192-49-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/200192-49-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart