AMBH324A54B5 Ambeed, Inc.

Boc-Thr-OMe

  • MDL number MFCD00270479
  • Cas Number : 79479-07-5
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79479-07-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/79479-07-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart