AMBH324A54BD Ambeed, Inc.

(R)-Methyl 3-((tert-butoxycarbonyl)amino)butanoate

  • MDL number MFCD03788463
  • Cas Number : 159877-47-1
  • Molecular Weight : 217.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/159877-47-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/159877-47-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart