AMBH324A54C0 Ambeed, Inc.

(S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide

  • MDL number MFCD02656449
  • Cas Number : 96293-17-3
  • Molecular Weight : 384.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/96293-17-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/96293-17-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart