AMBH324A54E0 Ambeed, Inc.

6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-7H-purine-8(9H)-thione

  • MDL number N/A
  • Cas Number : 3001-45-4
  • Molecular Weight : 299.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3001-45-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/3001-45-4.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart