AMBH324A54E1 Ambeed, Inc.

(S)-2-((S)-2-Amino-3-(1H-imidazol-4-yl)propanamido)-4-methylpentanoic acid

  • MDL number MFCD00037860
  • Cas Number : 7763-65-7
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/7763-65-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7763-65-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart