AMBH324A54E8 Ambeed, Inc.

4-Propyl-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

  • MDL number MFCD09750964
  • Cas Number : 122957-72-6
  • Molecular Weight : 320.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/122957-72-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/122957-72-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart