AMBH324A54EA Ambeed, Inc.

1,2-O-Isopropylidene-alpha-D-glucofuranose

  • MDL number MFCD00063244
  • Cas Number : 18549-40-1
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/18549-40-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/18549-40-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart