AMBH324A54EC Ambeed, Inc.

(trans,trans)-4-Pentyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)

  • MDL number MFCD17019208
  • Cas Number : 137644-54-3
  • Molecular Weight : 366.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/137644-54-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/137644-54-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart