AMBH324A54F4 Ambeed, Inc.

Rosuvastatin Calcium

  • MDL number MFCD04112702
  • Cas Number : 147098-20-2
  • Molecular Weight : 1001.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/147098-20-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/147098-20-2.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart