AMBH324A5501 Ambeed, Inc.

(S)-2-(2-Aminoacetamido)-4-methylpentanoic acid

  • MDL number MFCD00008127
  • Cas Number : 869-19-2
  • Molecular Weight : 188.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/869-19-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/869-19-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart