AMBH324A550A Ambeed, Inc.

N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide

  • MDL number MFCD03411210
  • Cas Number : 122833-58-3
  • Molecular Weight : 270.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/122833-58-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/122833-58-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart