AMBH324A5511 Ambeed, Inc.

(S)-tert-Butyl (1-cyclohexyl-2-hydroxyethyl)carbamate

  • MDL number MFCD04112591
  • Cas Number : 107202-39-1
  • Molecular Weight : 243.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/107202-39-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/107202-39-1.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart