AMBH324A5521 Ambeed, Inc.

Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-4-methoxy-4-oxobutanoate

  • MDL number MFCD00270494
  • Cas Number : 135941-84-3
  • Molecular Weight : 428.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/135941-84-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/135941-84-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart