AMBH324A5523 Ambeed, Inc.

(trans-4-Methylpyrrolidin-3-yl)methanol

  • MDL number MFCD10001415
  • Cas Number : 945723-36-4
  • Molecular Weight : 115.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/945723-36-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/945723-36-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart