AMBH324A5538 Ambeed, Inc.

Fmoc-D-Lys(Fmoc)-OH

  • MDL number MFCD00270743
  • Cas Number : 75932-02-4
  • Molecular Weight : 590.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/75932-02-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/75932-02-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart