AMBH324A553F Ambeed, Inc.

(R)-Benzyl (1-oxo-3-phenylpropan-2-yl)carbamate

  • MDL number MFCD01317841
  • Cas Number : 63219-70-5
  • Molecular Weight : 283.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/63219-70-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/63219-70-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart