AMBH324A5552 Ambeed, Inc.

Fmoc-Ser(Ac)-OH

  • MDL number MFCD00077070
  • Cas Number : 171778-17-9
  • Molecular Weight : 369.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/171778-17-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/171778-17-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart