AMBH324A5553 Ambeed, Inc.

Fmoc-Lys-OAll.HCl

  • MDL number MFCD08274506
  • Cas Number : 815619-80-8
  • Molecular Weight : 444.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/815619-80-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/815619-80-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart