AMBH324A5554 Ambeed, Inc.

Boc-Cys(Dpm)-OH

  • MDL number MFCD00151869
  • Cas Number : 21947-97-7
  • Molecular Weight : 387.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21947-97-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21947-97-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart