AMBH324A555F Ambeed, Inc.

(S)-Methyl 2-(ethylamino)-3-methylbutanoate

  • MDL number MFCD12172442
  • Cas Number : 222550-60-9
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/222550-60-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/222550-60-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart