AMBH324A557F Ambeed, Inc.

H-Cys(tBu)-OH.HCl

  • MDL number MFCD00037426
  • Cas Number : 2481-09-6
  • Molecular Weight : 213.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2481-09-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2481-09-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart