AMBH324A55A6 Ambeed, Inc.

Inosine 5'-monophosphate disodium salt hydrate

  • MDL number MFCD00150372
  • Cas Number : 352195-40-5
  • Molecular Weight : 410.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/352195-40-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/352195-40-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart