AMBH324A55AF Ambeed, Inc.

Fmoc-Cys(4-MeBzl)-OH

  • MDL number MFCD00065634
  • Cas Number : 136050-67-4
  • Molecular Weight : 447.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/136050-67-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/136050-67-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart