AMBH324A55B3 Ambeed, Inc.

(S)-2-(((Benzyloxy)carbonyl)amino)-4-hydroxybutanoic acid

  • MDL number MFCD00056718
  • Cas Number : 35677-88-4
  • Molecular Weight : 253.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/35677-88-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/35677-88-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart