AMBH324A55B4 Ambeed, Inc.

(R)-1-(Oxazol-2-yl)-2-phenylethanamine hydrochloride

  • MDL number MFCD30489020
  • Cas Number : 2044711-31-9
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2044711-31-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2044711-31-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart