AMBH324A55BF Ambeed, Inc.

(R)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid

  • MDL number MFCD02094549
  • Cas Number : 127474-54-8
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/127474-54-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/127474-54-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart