AMBH324A55D4 Ambeed, Inc.

(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride

  • MDL number MFCD11101236
  • Cas Number : 1251903-83-9
  • Molecular Weight : 280.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1251903-83-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1251903-83-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart