AMBH324A55DD Ambeed, Inc.

BIX-02188(Random Configuration)

  • MDL number MFCD14584538
  • Cas Number : 1094614-84-2
  • Molecular Weight : 412.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1094614-84-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1094614-84-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart