AMBH324A55E0 Ambeed, Inc.

Z-Glu-OtBu

  • MDL number MFCD00077016
  • Cas Number : 5891-45-2
  • Molecular Weight : 337.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5891-45-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5891-45-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart