AMBH324A55E7 Ambeed, Inc.

(R)-2-(2-(tert-Butoxy)-2-oxoethyl)-4-methylpentanoic acid

  • MDL number MFCD24396363
  • Cas Number : 112245-04-2
  • Molecular Weight : 230.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/112245-04-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112245-04-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart