AMBH324A55F1 Ambeed, Inc.

trans-4-Pentylcyclohexanecarboxylic acid

  • MDL number MFCD00009964
  • Cas Number : 38289-29-1
  • Molecular Weight : 198.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/38289-29-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/38289-29-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart