AMBH324A55F8 Ambeed, Inc.

Diammonium glycyrrhizinate

  • MDL number MFCD08460395
  • Cas Number : 79165-06-3
  • Molecular Weight : 856.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79165-06-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/79165-06-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart