AMBH324A55FB Ambeed, Inc.

(3S,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-6-methylheptanoic acid

  • MDL number MFCD0006567
  • Cas Number : 158257-40-0
  • Molecular Weight : 397.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/158257-40-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/158257-40-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart